Merck
  • Phenyliodine bis(trifluoroacetate)-mediated oxidative C-C bond formation: synthesis of 3-hydroxy-2-oxindoles and spirooxindoles from anilides.
Phenyliodine bis(trifluoroacetate)-mediated oxidative C-C bond formation: synthesis of 3-hydroxy-2-oxindoles and spirooxindoles from anilides.
Organic letters (2012-04-14)
Junwei Wang, Yucheng Yuan, Rui Xiong, Daisy Zhang-Negrerie, Yunfei Du, Kang Zhao
ABSTRACT
The reaction of phenyliodine bis(trifluoroacetate) (PIFA) with a series of anilides 1 (E = CO(2)Et) in CF(3)CH(2)OH was found to give 3-hydroxy-2-oxindole derivatives 2, while that with various anilides 1' (E = CON(R(4))Ar) afforded the C(2)-symmetric or unsymmetric spirooxindoles 3. These processes feature a metal-free oxidative C(sp(2))-C(sp(3)) bond formation, followed by oxidative hydroxylation or spirocyclization.
Sigma-Aldrich
2-Oxindole, 97%