Merck
  • Anthogorgienes A-O, new guaiazulene-derived terpenoids from a Chinese gorgonian Anthogorgia species, and their antifouling and antibiotic activities.
Anthogorgienes A-O, new guaiazulene-derived terpenoids from a Chinese gorgonian Anthogorgia species, and their antifouling and antibiotic activities.
Journal of agricultural and food chemistry (2011-12-14)
Dawei Chen, Shanjiang Yu, Leen van Ofwegen, Peter Proksch, Wenhan Lin
ABSTRACT
Fifteen new guaiazulene-based terpenoids designated anthogorgienes A-O (1-15) were isolated from a Chinese gorgonian Anthogorgia sp., together with eight known analogues (16-23). The structural patterns were classified into monomers, dimers, and trimers, which were supposed to be generated from a precursor guaiazulene and followed by side-chain and nucleus oxidation and oxidative rearrangement. The structures of new compounds were elucidated on the basis of extensive spectroscopic (IR, MS, and 1D and 2D NMR) data analysis. A possible biogenetic relationship of the isolated compounds was postulated. Some of the compounds showed potent antifouling activities against the larval settlement of barnacle Balanus amphitrite , whereas their antibiotic activities were also evaluated.
Sigma-Aldrich
Guaiazulene, 99%