MilliporeSigma
  • Standardization of hyphal growth inhibition rate as a means of evaluating Microsporum spp. in vitro susceptibility to terbinafine, griseofulvin, and ciclopiroxolamine.
Standardization of hyphal growth inhibition rate as a means of evaluating Microsporum spp. in vitro susceptibility to terbinafine, griseofulvin, and ciclopiroxolamine.
Mycopathologia (2011-05-26)
Fernanda Simas Corrêa Biancalana, Luzia Lyra, Maria Luiza Moretti, Katsuhiko Kamei, Angélica Zaninelli Schreiber
ABSTRACT
Reference methods for antifungal susceptibility tests recommend the use of conidia as inoculum. However, some isolates produce few conidia, while the invasive form of filamentous fungi in general is hyphae making susceptibility tests infeaseble. These facts suggest that other than conidia broth dilution method is required for susceptibility tests. The aim of this study was to clarify if the hyphal growth inhibition rate could be used as a method of determining the antifungal susceptibility of genus Microsporum. For this reason, a method which traces hyphal tips automatically and measures their growth rate was standardized for Microsporum spp. Control growth curves and test growth curves obtained by real-time observation of the hyphae groups responses to different concentrations of terbinafine, griseofulvin, and ciclopiroxolamine were used to compare with minimum inhibitory concentrations (MICs) obtained by conidia broth microdilution method. A visible reduction in the growth inhibition rate was observed when hyphal activity was evaluated using the third or fourth serial two-fold dilution below the MIC determined by broth microdilution for terbinafine and ciclopiroxolamine. For griseofulvin, this reduction occurred after the fifth dilution below the MIC. This study highlights the importance of the inoculum type used to determine the in vitro susceptibility of Microsporum strains. We conclude that measurement of hyphal growth inhibition, despite being time consuming, could be a suitable method for evaluating antifungal susceptibility, particularly for fungi as Microsporum spp. that produce a small (or not at all) number of conidia.
Product Description
Ciclopirox, European Pharmacopoeia (EP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
Ciclopirox olamine, European Pharmacopoeia (EP) Reference Standard