Merck
  • Impedance analysis of complex formation equilibria in phosphatidylcholine bilayers containing decanoic acid or decylamine.
Impedance analysis of complex formation equilibria in phosphatidylcholine bilayers containing decanoic acid or decylamine.
Cell biochemistry and biophysics (2011-02-23)
Monika Naumowicz, Aneta Dorota Petelska, Zbigniew Artur Figaszewski
ABSTRACT
Bilayer lipid membranes composed of phosphatidylcholine and decanoic acid or phosphatidylcholine and decylamine were investigated using electrochemical impedance spectroscopy. Interaction between membrane components causes significant deviations from the additivity rule. Area, capacitance, and stability constant values for the complexes were calculated based on the model assuming 1:1 stoichiometry, and the model was validated by comparison of these values to experimental results. We established that phosphatidylcholine and decylamine form highly stable 1:1 complexes. In the case of decanoic acid-modified phosphatidylcholine membranes, complexes with stoichiometries other than 1:1 should be taken into consideration.
Sigma-Aldrich
Decylamine, 95%
Sigma-Aldrich
Decylamine, ≥99.0% (GC)