Merck
N-phthaloyl-glycine-hydroxamic acid as serum iron chelator in rats.
Acta pharmaceutica (Zagreb, Croatia) (2008-06-03)
Julija Matijević-Sosa, Ita Samarzija, Lorena Honović, Blazenka Jurisić
ABSTRACT
The aim of this study was to investigate the activity of N-phthaloyl-glycine-hydroxamic acid (Phth-Gly-HA) as a new iron chelator in vivo to be used in iron overload diseases. After intraperitoneal application of Phth-Gly-HA to male rats (1 mg kg(-1) body mass) once a day for seven days, iron serum level decreased by 21%, whereas the iron value dropped by 32% in female rats (1.5 mg kg(-1) body mass). The results indicate that the tested substance has the ability to bind serum iron by complexation. Besides transferrin iron release, mobilization of ferritin iron is also possible.
Sigma-Aldrich
N-Phthaloylglycine, 97%