Merck
  • Synthesis, conformational analysis, and biological evaluation of 1-hexylindolactam-V10 as a selective activator for novel protein kinase C isozymes.
Synthesis, conformational analysis, and biological evaluation of 1-hexylindolactam-V10 as a selective activator for novel protein kinase C isozymes.
Journal of medicinal chemistry (2007-12-13)
Ryo C Yanagita, Yu Nakagawa, Nobuhiro Yamanaka, Kaori Kashiwagi, Naoaki Saito, Kazuhiro Irie
ABSTRACT
Conventional and novel protein kinase C (PKC) isozymes are the main targets of tumor promoters. We developed 1-hexylindolactam-V10 ( 5) as a selective activator for novel PKC isozymes that play important roles in various cellular processes related to tumor promotion, ischemia--reperfusion injury in the heart, and Alzheimer's disease. The compound existed as a mixture of three conformers. The trans-amide restricted analogues of 5 ( 14 and 15) hardly bound to PKC isozymes, suggesting that the active conformation of 5 could be that with a cis-amide. Compound 5 selectively translocated novel PKC isozymes over conventional PKC isozymes in HeLa cells at 0.1-1 microM. These results suggest that 5 could be useful for the functional analysis of novel PKC isozymes.
Sigma-Aldrich
(−)-Indolactam V, ≥96% (HPLC)