Merck
[A case of methemoglobinemia due to 4-nitrobenzonitrile exposure].
La Medicina del lavoro (1991-03-01)
G Monechi, G Fabrizi de Biani, L Colasanti
ABSTRACT
The paper describes a singular case of acute intoxication by 4-nitrobenzonitrile, a rare intermediate compound produced by pharmaceutical synthesis. The patients subsequently showed an increase in triglycerides and fatty alterations of the liver.
Sigma-Aldrich
4-Nitrobenzonitrile, 97%