Merck
  • Joint effects of microwave and chromium trioxide on root tip cells of Vicia faba.
Joint effects of microwave and chromium trioxide on root tip cells of Vicia faba.
Journal of Zhejiang University. Science. B (2006-02-28)
Xiao-wei Qian, Wei-hua Luo, Ou-xiang Zheng
ABSTRACT
The mutagenic effects of microwave and chromium trioxide (CrO(3)) on Vicia faba root tip were studied. Micronucleus assay and chromosomal aberration assay were used to determine the mitotic index, the micronucleus frequency and chromosomal aberration frequency of Vicia faba root tip cells induced by microwave and CrO(3). The results showed that the micronucleus frequency decreased, and that the mitotic index and chromosomal aberration frequency showed linear dose responses to CrO(3), in treatment of microwave for 5 s. In microwave of 25 s, the mitotic index decreased, the micronucleus frequency and chromosomal aberration frequency increased with increase of CrO(3) concentration. We concluded that microwave and CrO(3) had antagonistic effect on the mitotic index of Vicia faba root tip cells, but had synergetic effect on micronucleus frequency and chromosomal aberration frequency of Vicia faba root tip cells.
Sigma-Aldrich
Chromium(VI) oxide, 99.99% trace metals basis
Sigma-Aldrich
Chromium(VI) oxide, ACS reagent, ≥98.0%
Sigma-Aldrich
Chromium(VI) oxide, ReagentPlus®, 99.9% trace metals basis