Merck
  • Caspase-independent cell death by arsenic trioxide in human cervical cancer cells: reactive oxygen species-mediated poly(ADP-ribose) polymerase-1 activation signals apoptosis-inducing factor release from mitochondria.
Caspase-independent cell death by arsenic trioxide in human cervical cancer cells: reactive oxygen species-mediated poly(ADP-ribose) polymerase-1 activation signals apoptosis-inducing factor release from mitochondria.
Cancer research (2004-12-18)
Young-Hee Kang, Min-Jung Yi, Min-Jung Kim, Moon-Taek Park, Sangwoo Bae, Chang-Mo Kang, Chul-Koo Cho, In-Chul Park, Myung-Jin Park, Chang Hun Rhee, Seok-Il Hong, Hee Yong Chung, Yun-Sil Lee, Su-Jae Lee
ABSTRACT
Although mechanisms of arsenic trioxide (As(2)O(3))-induced cell death have been studied extensively in hematologic cancers, those in solid cancers have yet to be clearly defined. In this study, we showed that the translocation of apoptosis-inducing factor (AIF) from mitochondria to the nucleus is required for As(2)O(3)-induced cell death in human cervical cancer cells. We also showed that reactive oxygen species (ROS)-mediated poly(ADP-ribose) polymerase-1 (PARP-1) activation is necessary for AIF release from mitochondria. The treatment of human cervical cancer cells with As(2)O(3) induces dissipation of mitochondrial membrane potential (Deltapsi(m)), translocation of AIF from mitochondria to the nucleus, and subsequent cell death. Small interfering RNA targeting of AIF effectively protects cervical cancer cells against As(2)O(3)-induced cell death. As(2)O(3) also induces an increase of intracellular ROS level and a marked activation of PARP-1. N-acetyl-l-cystein, a thiol-containing antioxidant, completely blocks As(2)O(3)-induced PARP-1 activation, Deltapsi(m) loss, nuclear translocation of AIF from mitochondria, and the consequent cell death. Furthermore, pretreatment of 1,5-dihydroxyisoquinoline or 3,4-dihydro-5-[4-(1-piperidinyl)butoxy]-1(2H)-isoquinolinone, PARP-1 inhibitors, effectively attenuates the loss of Deltapsi(m), AIF release, and cell death. These data support a notion that ROS-mediated PARP-1 activation signals AIF release from mitochondria, resulting in activation of a caspase-independent pathway of cell death in solid tumor cells by As(2)O(3) treatment.
Sigma-Aldrich
Arsenic(III) chloride, 99.99% trace metals basis