Merck
Synthesis of radioiodinated aryl iodides via boronate precursors.
Nuclear medicine and biology (2002-11-28)
George W Kabalka, Murthy R Akula, Jianhua Zhang
ABSTRACT
Arylboronate esters are converted to iodine-123 labeled aryl iodides using no-carrier-added iodine-123 labeled sodium iodide in the presence of chloramine-T. High yields of radiochemically pure products are obtained.
Sigma-Aldrich
4-Iodoanisole, 98%