Merck
  • Initial study in rats evaluating the effects of 1,4-dichlorobutene-2 (DCB) on the respiratory tract.
Initial study in rats evaluating the effects of 1,4-dichlorobutene-2 (DCB) on the respiratory tract.
Drug and chemical toxicology (2002-05-25)
L S Mullin, T Chiu, G L Kennedy
ABSTRACT
Rats were exposed by inhalation to either 0.5 ppm 1,4-dichlorobutene-2 (DCB) for two years or to 5.0 ppm for seven months, 2.5 ppm for five months, and no further exposure for 12 months prior to sacrifice. Malignant and non-malignant tumors of the nasal tissues were seen in both test groups with the incidence and proportion of malignant tumors being much higher in the 5.0/2.5 ppm rats. Under the conditions of this study, DCB is carcinogenic in rats of both sexes.
Sigma-Aldrich
trans-1,4-Dichloro-2-butene, 98%
Sigma-Aldrich
trans-1,4-Dichloro-2-butene, technical grade, 85%, remainder predominantly cis isomer
Sigma-Aldrich
cis-1,4-Dichloro-2-butene, 95%