Merck
  • Crystal structures of mouse class II alcohol dehydrogenase reveal determinants of substrate specificity and catalytic efficiency.
Crystal structures of mouse class II alcohol dehydrogenase reveal determinants of substrate specificity and catalytic efficiency.
Journal of molecular biology (2000-09-06)
S Svensson, J O Höög, G Schneider, T Sandalova
ABSTRACT
The structure of mouse class II alcohol dehydrogenase (ADH2) has been determined in a binary complex with the coenzyme NADH and in a ternary complex with both NADH and the inhibitor N-cyclohexylformamide to 2.2 A and 2.1 A resolution, respectively. The ADH2 dimer is asymmetric in the crystal with different orientations of the catalytic domains relative to the coenzyme-binding domains in the two subunits, resulting in a slightly different closure of the active-site cleft. Both conformations are about half way between the open apo structure and the closed holo structure of horse ADH1, thus resembling that of ADH3. The semi-open conformation and structural differences around the active-site cleft contribute to a substantially different substrate-binding pocket architecture as compared to other classes of alcohol dehydrogenase, and provide the structural basis for recognition and selectivity of alcohols and quinones. The active-site cleft is more voluminous than that of ADH1 but not as open and funnel-shaped as that of ADH3. The loop with residues 296-301 from the coenzyme-binding domain is short, thus opening up the pocket towards the coenzyme. On the opposite side, the loop with residues 114-121 stretches out over the inter-domain cleft. A cavity is formed below this loop and adds an appendix to the substrate-binding pocket. Asp301 is positioned at the entrance of the pocket and may control the binding of omega-hydroxy fatty acids, which act as inhibitors rather than substrates. Mouse ADH2 is known as an inefficient ADH with a slow hydrogen-transfer step. By replacing Pro47 with His, the alcohol dehydrogenase activity is restored. Here, the structure of this P47H mutant was determined in complex with NADH to 2.5 A resolution. His47 is suitably positioned to act as a catalytic base in the deprotonation of the substrate. Moreover, in the more closed subunit, the coenzyme is allowed a position closer to the catalytic zinc. This is consistent with hydrogen transfer from an alcoholate intermediate where the Pro/His replacement focuses on the function of the enzyme.
Sigma-Aldrich
12-Hydroxydodecanoic acid, 97%
Sigma-Aldrich
N-Cyclohexylformamide, 99%