Merck
  • Effects of mimosine and 2,3-dihydroxypyridine on fiber shedding in Angora goats.
Effects of mimosine and 2,3-dihydroxypyridine on fiber shedding in Angora goats.
Journal of animal science (1999-05-26)
P J Reis, R Puchala, T Sahlu, A L Goetsch
ABSTRACT
The effects of intravenous infusion of mimosine or 2,3-dihydroxypyridine (2,3-DHP) and the effects of oral dose level of mimosine on fiber shedding in Angora goats were determined. In one experiment, 20 mature Angora wethers (36+/-1.9 kg BW) were infused for 2 d with 79, 102, or 135 mg/(kg BW.d) of mimosine, 90 mg/(kg BW.d) of 2,3-DHP, or saline. At 7 d after infusion began, fiber shedding was observed in all goats receiving mimosine but not in any goats infused with 2,3-DHP or saline. Fiber shedding varied among goats; in some goats, fiber shedding was complete and occurred without hand-plucking, whereas in others fiber was retained by nonshed fibers but could be removed by hand-plucking. Nonshed fibers were larger in diameter and more likely to be medullated (P < .05) compared with hand-plucked fibers. Mean plasma mimosine concentration at 24 and 48 h after infusion began was 79 and 98 micromol/L (P < .05), respectively, and greater (P < .05) for mimosine infused at 135 than at 102 mg/(kg BW.d) (89, 68, and 108 micromol/L for mimosine infused at 79, 102, and 135 mg/[kg BW.d], respectively; SE 9.5). In another experiment, oral dosing of eight Angora bucks (23+/-.5 kg BW) with 400 or 600 mg/kg BW of mimosine rapidly increased plasma mimosine concentration, which reached approximately 100 and 160 micromol/L at 5 h after dosing; however, periods of time during which plasma mimosine concentrations were comparable to those in the first experiment were considerably shorter. Oral mimosine dosing did not induce fiber shedding in 7 d. After 31 d, fiber was retained by nonshed fibers but could be removed by hand-plucking or could only be partially removed with difficulty by hand-plucking. There were no toxic effects of mimosine or 2,3-DHP administration; only minor, short-term inhibitions of feed intake by mimosine were noted in some goats. In conclusion, mimosine holds promise as a safe means to remove fiber of Angora goats; further research is necessary to characterize the seasonality of follicle activity and to develop convenient means of mimosine delivery.
Sigma-Aldrich
2,3-Dihydroxypyridine, 95%