Merck
  • Pharmacological comparison of the effect of ibogaine and 18-methoxycoronaridine on isolated smooth muscle from the rat and guinea-pig.
Pharmacological comparison of the effect of ibogaine and 18-methoxycoronaridine on isolated smooth muscle from the rat and guinea-pig.
British journal of pharmacology (2000-04-26)
M K Mundey, N A Blaylock, R Mason, S D Glick, I M Maisonneuve, V G Wilson
ABSTRACT
Ibogaine and 18-methoxycoronaridine are naturally occurring alkaloids reported to possess antiaddictive properties in several models of drug dependence. We have examined their effect at mu-opioid receptors regulating neurogenic contractions of several smooth muscle preparations and also against spontaneous contractions of the rat isolated portal vein. Ibogaine (pIC(50) 5.28) and 18-methoxycoronaridine (pIC(50) 5.05) caused a concentration-dependent inhibition of cholinergic contractions of the guinea-pig ileum which was not affected by the opioid receptor antagonist naloxone (1 microM). In the rat isolated vas deferens ibogaine and 18-methoxycoronaridine caused a concentration-dependent enhancement of purinergic contractions. Both agents (30 microM) caused a 3 - 5 fold rightward displacement of DAMGO-induced inhibition of purinergic contractions, but similar effects were observed for ibogaine against alpha(2)-adrenoceptor-mediated inhibition of neurogenic responses. In the guinea-pig isolated bladder both ibogaine (10 microM) and 18-methoxycoronaridine (10 microM) caused a 2 fold increase in the purinergic component of neurogenic contractions without significantly altering cholinergic contractions or responses to exogenous ATP. In contrast, ibogaine (1 - 30 microM), but not 18-methoxycoronaridine, caused a concentration-dependent enhancement of spontaneous contractions of the rat isolated portal vein. In summary, while ibogaine and 18-methoxycoronaridine modulated electrically-evoked contractions in the three preparations examined, we have no evidence for a selective interaction with pre-junctional mu-opioid receptors. The pronounced enhancement of purinergic contractions produced by both agents is a novel finding and worthy of further investigation.