Merck
  • Discovery and design of novel and selective vasopressin and oxytocin agonists and antagonists: the role of bioassays.
Discovery and design of novel and selective vasopressin and oxytocin agonists and antagonists: the role of bioassays.
Experimental physiology (2000-05-05)
W Y Chan, N C Wo, S T Stoev, L L Cheng, M Manning
ABSTRACT
Synthetic oxytocin and vasopressin agonists and antagonists have become important tools for research and were instrumental in the identification of the four known receptor subtypes, V1a, V2, V1b (V3) and oxytocin, of these peptide hormones. However, the relative lack of receptor selectivity, particularly of the antagonists, has limited their usefulness as experimental probes and their potential as therapeutic agents. We now present some findings from our continuing studies aimed at the design of more selective oxytocin and vasopressin agonists and antagonists and a structure-activity relationship update on our recently discovered novel hypotensive vasopressin peptides. Bioassays have been, and continue to be, of critical importance in leading to the discovery of the novel agonists, antagonists and hypotensive peptides reported here. This paper highlights three main aspects of these studies. (1) Replacement of the tyrosine2 and/or phenylalanine3 residues in the V2 agonist deamino,[Val4,D-Arg8]arginine-vasopressin (dVDAVP) by thienylalanine resulted in selective V2 agonists with strikingly high potencies. However, the peptide solutions were unstable and lost activity over time. These highly potent V2 agonists, which are devoid of vasopressor activity, are promising leads for improving drugs for treating diabetes insipidus, enuresis and coagulation disorders. (2) Diaminopropionic acid and diaminobutyric acid substitution at position-5 in oxytocin and in V1a antagonists yielded, respectively, the first specific antagonist for the oxytocin receptor, desGly-NH2,d(CH2)5[D-Trp2,Thr4,Dap5]OVT and the first specific antagonist for the vasopressin V1a receptor, d(CH2)5[Tyr(Me)2,Dab5]AVP. The availability of single receptor subtype-specific or selective antagonists will enhance our ability to delineate receptor functions. Utilising these new receptor specific probes, we were able to show that the uterotonic action of vasopressin is mediated principally by oxytocin and not by V1a receptors. (3) Replacement of the phenylalanine3 residue in the V1a/V2/oxytocin antagonist, d(CH2)5[D-Tyr(Et)2,Val4]AVP, with arginine3 yielded the novel, selective, hypotensive vasopressin peptide, d(CH2)5[D-Tyr(Et)2,Arg3,Val4]AVP (Peptide I). Bioassay characterisations of Peptide I show that its vasodepressor action is independent of the peripheral autonomic, bradykinin, nitric oxide and prostaglandin systems and is not mediated by the known classical oxytocin and vasopressin receptors. These findings suggest the existence of a new vasopressin receptor subtype that may be relevant to the vasodilating action of vasopressin in regional vascular beds. Iodinatable hypotensive peptides have been synthesised and could be developed as markers for the putative new receptor. Ongoing structure-activity relationship studies on Peptide I have led to more potent and selective hypotensive peptides for use as new research tools and as leads for the development of a new class of antihypertensive agents.
Sigma-Aldrich
[β-Mercapto-β,β-cyclopentamethylenepropionyl1, O-Et-Tyr2, Val4, Arg8]-Vasopressin, ≥97% (HPLC)