Merck
Identification of prostaglandin receptors in human ureters.
BMC urology (2012-12-12)
Matthias Oll, Claudia Baumann, Turang E Behbahani, Alexander von Ruecker, Stefan C Müller, Jörg Ellinger
ABSTRACT
Prostaglandins play an important role in ureteral obstruction, but the detailed expression profiles of the prostaglandin receptors (PTGER1, PTGER2, PTGER3, PTGER4, PTGFR) remain unknown in the different parts of the human ureter. The expression pattern of PTGER1, PTGER2, PTGER3, PTGER4 and PTGFR was determined in human distal, mid and proximal ureter and renal pelvis samples using immunohistochemistry (protein levels) and quantitative real-time PCR (mRNA). PTGER1 was highly expressed in most samples irrespective of the ureteral localization; however, urothelial cells had higher levels of PTGER1 than smooth muscle cells. PTGFR was also moderately to strongly expressed in urothelial and smooth muscle cells. In comparison, PTGER2-4 expression was mostly unexpressed or weakly expressed in urothelial and smooth cells in all regions. Our data indicate high levels of PTGER1 in ureters.
Sigma-Aldrich
Anti-FP Prostanoid Receptor antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution