Merck
  • Development of tight junction-strengthening compounds using a high-throughput screening system to evaluate cell surface-localized claudin-1 in keratinocytes.
Development of tight junction-strengthening compounds using a high-throughput screening system to evaluate cell surface-localized claudin-1 in keratinocytes.
Scientific reports (2024-02-09)
Hiroki Sakamoto, Momoyo Nishikawa, Seigo Yamada
ABSTRACT
Tight junctions (TJs) are important factors constituting the physical barriers of the skin, and their suppression has been described in various conditions, such as aged skin and atopic dermatitis lesions. However, the methods for improving skin TJ function remain insufficient. Therefore, to obtain compounds that can improve TJ function, we developed a novel high-throughput screening system termed live-cell immunostaining to evaluate cell surface-localized claudin-1 (CLDN1) with high selectivity using normal human epidermal keratinocytes (NHEKs). Heparinoid and phospho-pyridoxal (p-Pyr), a metabolite of pyridoxine, were identified as hit compounds. In addition, heparinoid was strongly suggested to increase CLDN1 expression by inhibiting epidermal growth factor receptor signaling. By contrast, p-Pyr did not enhance CLDN1 expression, but it accelerated the translocation of CLDN1 to the cell surface. Finally, we confirmed that heparinoid and p-Pyr improved barrier function in NHEKs in a transepithelial electrical resistance assay. In conclusion, heparinoid and p-Pyr could potentially ameliorate skin conditions by improving TJ function.
Sigma-Aldrich
Anti-Claudin-1/CLDN1 Antibody, clone 7A5, clone 7A5, from mouse