Merck
  • Lubiprostone is non-selective activator of cAMP-gated ion channels and Clc-2 has a minor role in its prosecretory effect in intestinal epithelial cells.
Lubiprostone is non-selective activator of cAMP-gated ion channels and Clc-2 has a minor role in its prosecretory effect in intestinal epithelial cells.
Molecular pharmacology (2022-06-10)
Apurva A Oak, Tifany Chu, Pattareeya Yottasan, Parth D Chhetri, Jie Zhu, Justin Du Bois, Onur Cil
ABSTRACT
Loss of prosecretory Cl- channel CFTR activity is considered as the key cause of gastrointestinal disorders in cystic fibrosis including constipation and meconium ileus. Clc-2 is proposed as an alternative Cl- channel in intestinal epithelia that can compensate for CFTR loss-of-function. Lubiprostone is an FDA-approved drug with Clc-2 activation as its presumed mechanism of action. However, relative contribution of Clc-2 in intestinal Cl- secretion and the mechanism of action of lubiprostone remain controversial due to lack of selective Clc-2 inhibitors. Using recently identified selective Clc-2 inhibitor AK-42, we characterized the roles of Clc-2 in Cl- secretion in human intestinal epithelial T84 cells. Clc-2 inhibitor AK-42 had minimal (15%) inhibitory effect on secretory short-circuit current (Isc) induced by cAMP agonists, where subsequently applied CFTR inhibitor (CFTRinh-172) caused 2-3 fold greater inhibition. Similarly, AK-42 inhibited lubiprostone-induced secretory Isc by 20%, whereas CFTRinh-172 caused 2-3 fold greater inhibition. In addition to increasing CFTR and Clc-2-mediated apical Cl- conductance, lubiprostone increased basolateral membrane K+ conductance, which was completely reversed by cAMP-activated K+ channel inhibitor BaCl2 All components of lubiprostone-induced secretion (Clc-2, CFTR and K+ channels) were inhibited by ~65% with the extracellular Ca2+-sensing receptor (CaSR) activator cinacalcet that stimulates cAMP hydrolysis. Lastly, EP4 prostaglandin receptor inhibitor GW627368 pretreatment inhibited lubiprostone-induced secretion by 40% without any effect on forskolin response. Our findings suggest that Clc-2 has minor role in cAMP-induced intestinal Cl- secretion; and lubiprostone is not a selective Clc-2 activator, but general activator of cAMP-gated ion channels in human intestinal epithelial cells. Significance Statement Cl- channel Clc-2 activation is the proposed mechanism of action of the FDA-approved constipation drug lubiprostone. Using first-in-class selective Clc-2 inhibitor AK-42, we showed that Clc-2 has minor contribution in intestinal Cl- secretion induced by lubiprostone and cAMP agonists. We also found that lubiprostone is a general activator of cAMP-gated ion channels in human intestinal epithelial cells (via EP4 receptors). Our findings clarify the roles of Clc-2 in intestinal Cl- secretion and elucidate the mechanism of action of approved-drug lubiprostone.
Sigma-Aldrich
CLC-2 channel inhibitor, AK-42, ≥98% (HPLC)