Merck
Colesevelam in the management of type 2 diabetes.
JPMA. The Journal of the Pakistan Medical Association (2020-05-14)
Sanjay Kalra, Gagan Priya, Sameer Aggarwal
ABSTRACT
Colesevelam is a bile acid sequestrant, approved for the management of both dyslipidaemia and type 2 diabetes. This review discusses the potential for the use of colesevelam in the management of type 2 diabetes. Expert opinion suggests possible indications where colesevelam may add value as a glucose lowering agent. It also highlights the limitations of the drug, and precautions that must be observed while using it.
Sigma-Aldrich
Colesevelam hydrochloride, ≥98% (HPLC)