- Comprehensive analysis of telomerase inhibition by gallotannin.
Comprehensive analysis of telomerase inhibition by gallotannin.
Oncotarget (2018-05-04)
Nikita Savelyev, Polina Baykuzina, Svetlana Dokudovskaya, Olga Lavrik, Maria Rubtsova, Olga Dontsova
PMID29721155
ABSTRACT
Gallotannin (or tannic acid) is a naturally occurring compound that inhibits cell growth and activity of different DNA-polymerases, including telomerase. The purpose of the present study was to gain insight into the mechanism of telomerase inhibition by gallotannin. We determined that gallotannin inhibits telomerase in vitro with an half maximal inhibitory concentrations value of 130 nM, but it does not affect telomerase complex assembly and component levels in vivo. The inhibitory activity of gallotannin against telomerase provides an additional explanation for the anti-cancer activities of this compound.
Sigma-Aldrich
Protein G Agarose/Salmon Sperm DNA, 2.5 mL, for use in chromatin immunoprecipitations (ChIP assays)