Merck
  • Host Interferon-Stimulated Gene 20 Inhibits Pseudorabies Virus Proliferation.
Host Interferon-Stimulated Gene 20 Inhibits Pseudorabies Virus Proliferation.
Virologica Sinica (2021-04-09)
Xiaoyong Chen, Dage Sun, Sujie Dong, Huanjie Zhai, Ning Kong, Hao Zheng, Wu Tong, Guoxin Li, Tongling Shan, Guangzhi Tong
ABSTRACT
Host interferon-stimulated gene 20 (ISG20) exerts antiviral effects on viruses by degrading viral RNA or by enhancing IFN signaling. Here, we examined the role of ISG20 during pseudorabies virus (PRV) proliferation. We found that ISG20 modulates PRV replication by enhancing IFN signaling. Further, ISG20 expression was upregulated following PRV infection and poly(I:C) treatment. Ectopic expression of ISG20 inhibited PRV proliferation in PK15 cells, whereas knockdown of ISG20 promoted PRV proliferation. In addition, ISG20 expression upregulated IFN-β expression and enhanced IFN downstream signaling during PRV infection. Notably, PRV UL24 suppressed the transcription of ISG20, thus antagonizing its antiviral effect. Further domain mapping analysis showed that the N terminus (amino acids 1-90) of UL24 was responsible for the inhibition of ISG20 transcription. Collectively, these findings characterize the role of ISG20 in suppressing PRV replication and increase the understanding of host-PRV interplay.
Sigma-Aldrich
Anti-MYC, affinity isolated antibody