Merck
  • Clec12a is an inhibitory receptor for uric acid crystals that regulates inflammation in response to cell death.
Clec12a is an inhibitory receptor for uric acid crystals that regulates inflammation in response to cell death.
Immunity (2014-03-19)
Konstantin Neumann, Mercedes Castiñeiras-Vilariño, Ulrike Höckendorf, Nicole Hannesschläger, Simone Lemeer, Danny Kupka, Svenia Meyermann, Maciej Lech, Hans-Joachim Anders, Bernhard Kuster, Dirk H Busch, Andreas Gewies, Ronald Naumann, Olaf Groß, Jürgen Ruland
ABSTRACT
Recognition of cell death by the innate immune system triggers inflammatory responses. However, how these reactions are regulated is not well understood. Here, we identify the inhibitory C-type lectin receptor Clec12a as a specific receptor for dead cells. Both human and mouse Clec12a could physically sense uric acid crystals (monosodium urate, MSU), which are key danger signals for cell-death-induced immunity. Clec12a inhibited inflammatory responses to MSU in vitro, and Clec12a-deficient mice exhibited hyperinflammatory responses after being challenged with MSU or necrotic cells and after radiation-induced thymocyte killing in vivo. Thus, we identified a negative regulatory MSU receptor that controls noninfectious inflammation in response to cell death that has implications for autoimmunity and inflammatory disease.
Sigma-Aldrich
Uric acid, BioXtra, ≥99% (HPLC)