Merck
  • Residues and dissipation kinetic of abamectin, chlorfenapyr and pyridaben acaricides in green beans (Phaseolus vulgaris L.) under field conditions using QuEChERS method and HPLC.
Residues and dissipation kinetic of abamectin, chlorfenapyr and pyridaben acaricides in green beans (Phaseolus vulgaris L.) under field conditions using QuEChERS method and HPLC.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes (2020-02-18)
Mohamed E I Badawy, Mostafa S Mahmoud, Marium M Khattab
ABSTRACT
The current study estimated the dissipation rates of abamectin, chlorfenapyr and pyridaben acaricides in pods of green beans (Phaseolus vulgaris L.) under field conditions in Egypt. Pesticides were extracted and cleaned-up by QuEChERS method and were analyzed by HPLC. The dissipation of these acaricides followed the first order kinetics model with half-life (t1/2) values 1.00, 3.50 and 1.50 days for abamectin, chlorfenapyr and pyridaben, respectively. The lowest residues, at different time intervals of field application rate of each pesticide, were observed with abamectin followed by pyridaben and then chlorfenapyr. Pre-harvest intervals (PHIs) were 10.00, 13.50 and 6.00 days for abamectin, chlorfenapyr and pyridaben, respectively and were below the established European maximum residue limits (EU MRLs) 10-14, 14-21 and 7-10 days after application, respectively. If the fresh pods will be consumed after harvest, it is expected that the presence of these pesticides in the food will have a negative impact on human health. Therefore, the elimination of the residues of these harmful pesticides must be carried out.