Merck
  • Non-targeted, high resolution mass spectrometry strategy for simultaneous monitoring of xenobiotics and endogenous compounds in green sea turtles on the Great Barrier Reef.
Non-targeted, high resolution mass spectrometry strategy for simultaneous monitoring of xenobiotics and endogenous compounds in green sea turtles on the Great Barrier Reef.
The Science of the total environment (2017-05-20)
Amy L Heffernan, Maria M Gómez-Ramos, Caroline Gaus, Soumini Vijayasarathy, Ian Bell, Christine Hof, Jochen F Mueller, Maria J Gómez-Ramos
ABSTRACT
Chemical contamination poses a threat to ecosystem, biota and human health, and identifying these hazards is a complex challenge. Traditional hazard identification relies on a priori-defined targets of limited chemical scope, and is generally inappropriate for exploratory studies such as explaining toxicological effects in environmental systems. Here we present a non-target high resolution mass spectrometry environmental monitoring study with multivariate statistical analysis to simultaneously detect biomarkers of exposure (e.g. xenobiotics) and biomarkers of effect in whole turtle blood. Borrowing the concept from clinical chemistry, a case-control sampling approach was used to investigate the potential influence of xenobiotics of anthropogenic origin on free-ranging green sea turtles (Chelonia mydas) from a remote, offshore 'control' site; and two coastal 'case' sites influenced by urban/industrial and agricultural activities, respectively, on the Great Barrier Reef in North Queensland, Australia. Multiple biomarkers of exposure, including sulfonic acids (n=9), a carbamate insecticide metabolite, and other industrial chemicals; and five biomarkers of effect (lipid peroxidation products), were detected in case sites. Additionally, two endogenous biomarkers of neuroinflammation and oxidative stress were identified, and showed moderate-to-strong correlations with clinical measures of inflammation and liver dysfunction. Our data filtering strategy overcomes limitations of traditional a priori selection of target compounds, and adds to the limited environmental xenobiotic metabolomics literature. To our knowledge this is the first case-control study of xenobiotics in marine megafauna, and demonstrates the utility of green sea turtles to link internal and external exposure, to explain potential toxicological effects in environmental systems.