Merck
  • 13C-labeled N-acetyl-neuraminic acid in aqueous solution: detection and quantification of acyclic keto, keto hydrate, and enol forms by 13C NMR spectroscopy.
13C-labeled N-acetyl-neuraminic acid in aqueous solution: detection and quantification of acyclic keto, keto hydrate, and enol forms by 13C NMR spectroscopy.
Journal of the American Chemical Society (2008-08-20)
Thomas Klepach, Ian Carmichael, Anthony S Serianni
ABSTRACT
Aqueous solutions of N-acetyl-neuraminic acid (Neu5Ac, 1) labeled with (13)C at C1, C2, and/or C3 were analyzed by (13)C NMR spectroscopy to detect and quantify the acyclic forms (keto, keto hydrate, enol) present at varying pHs. In addition to pyranoses, solutions contained the keto form, based on the detection of C2 signals at approximately 198 ppm (approximately 0.7% at pH 2). Spectra of [2-(13)C] and [3-(13)C] isotopomers contained signals arising from labeled carbons at approximately 143 and approximately 120 ppm, respectively, which were attributed to enol forms. Solution studies of [1,2,3-(13)C3] 1 substantiated the presence of enol (approximately 0.5% at pH 2). Enol was not detected at pH > 6.0. A C2 signal observed at approximately 94 ppm was identified as C2 of the keto hydrate (approximately 1.9% at pH 2), based partly on its abundance as a function of solution pH. Density functional theory (DFT) calculations were used to study the effect of enol and hydrate structure on J(CH) and J(CC) values involving C2 and C3 of these forms. Solvated DFT calculations showed that (2)J(C2,H3) in cis and trans enols have similar magnitudes but opposite signs, making this J-coupling potentially useful to distinguish enol configurations. Solvent deuterium exchange studies of 1 showed rapid incorporation of (2)H from (2)H2O at H3 axial in the pyranoses at p(2)H 8.0, followed by slower exchange at H3 equatorial. The acyclic keto form, which presumably participates in this reaction, must assume a pseudo-cyclic conformation in solution in order to account for the exchange selectivity. Weak (13)C signals arising from labeled species were also observed consistently and reproducibly in aqueous solutions of (13)C-labeled 1, possibly arising from products of lactonization or intermolecular esterification.
Sigma-Aldrich
N-Acetyl-D-neuraminic acid-1,2,3-13C3, ā‰„99 atom % 13C, ā‰„97% (CP)