Merck
  • Electrophysiological and Behavioral Responses of Dasychira baibarana (Lepidoptera: Lymantriidae) to Tea Plant Volatiles.
Electrophysiological and Behavioral Responses of Dasychira baibarana (Lepidoptera: Lymantriidae) to Tea Plant Volatiles.
Environmental entomology (2021-03-08)
Fida Hussain Magsi, Zongxiu Luo, Yingjie Zhao, Zhaoqun Li, Xiaoming Cai, Lei Bian, Zongmao Chen
ABSTRACT
Tea black tussock moth, Dasychira baibarana (Matsumura) (Lepidoptera: Lymantriidae), is a devastating pest species of the tea plant in China. Here, we evaluated the responses of D. baibarana to tea plant volatiles using gas chromatography coupled electroantennographic detection (GC-EAD), eleclectroantennography (EAG), and a Y-tube olfactometer. In total, 11 of 18 analyzed compounds elicited GC-EAD responses from test insects. GC-EAD bio-active compounds were further investigated using EAG and behavioral responses. In the EAG analysis, male moths had significantly greater responses to four compounds [(Z)-3-hexenyl butyrate, (Z)-3-hexen-1-ol, ocimene and benzyl alcohol] than female moths. For females, maximum EAG amplitudes, were recorded in response to linalool, (Z)-3-hexenyl hexanoate and (Z)-jasmone. In EAG and behavioral bio-assays, the responses of both sexes were dose independent. In behavioral bio-assays male moths responding significantly to (Z)-3-hexen-1-ol, ocimene, (Z)-3-hexenyl butyrate, linalool, benzyl alcohol, and (Z)-jasmone at various concentrations. For females, significant behavioral responses were observed to (Z)-3-hexenyl hexanoate, followed by (Z)-jasmone, linalool, ocimene, and benzyl alcohol. However, neither sex was sensitive to 4 of the 11 tested compounds, phenyethyl alcohol, phenylacetonitrile, (E)-nerolidol, and indole. The present results showed that tea plant volatiles influenced the behavior of D. baibarana moths, which will greatly contribute in developing eco-friendly control strategies for D. baibarana, through the application of a blend of compounds that showed significant EAG and behavioral responses or a blend combined with female-produced sex pheromones.