Merck
  • Synthesis and biological evaluation of technetium-labeled D-glucose-MAG3 derivative as agent for tumor diagnosis.
Synthesis and biological evaluation of technetium-labeled D-glucose-MAG3 derivative as agent for tumor diagnosis.
Bioorganic & medicinal chemistry letters (2009-04-01)
André Luís Branco de Barros, Valbert Nascimento Cardoso, Luciene das Graças Mota, Elaine Amaral Leite, Mônica Cristina de Oliveira, Ricardo José Alves
ABSTRACT
A d-glucose-MAG(3) derivative was successfully synthesized and radiolabeled in high labeling yield. Biodistribution studies in Ehrlich tumor-bearing mice were performed. This compound showed high accumulation in tumor tissue with high tumor-to-muscle ratio and moderate tumor-to-blood ratio. Thus, d-glucose-MAG(3) is a potential agent for tumor diagnosis.
Sigma-Aldrich
1-Azido-1-deoxy-β-D-glucopyranoside