- Expression of spindle assembly checkpoint proteins BubR1 and Mad2 expression as potential biomarkers of malignant transformation of oral leukoplakia: an observational cohort study.
Expression of spindle assembly checkpoint proteins BubR1 and Mad2 expression as potential biomarkers of malignant transformation of oral leukoplakia: an observational cohort study.
The Spindle Assembly Checkpoint (SAC) is a surveillance mechanism essential to ensure the accuracy of chromosome segregation during mitosis. Our aim was to evaluate the expression of SAC proteins in oral carcinogenesis, and to assess their potential in predicting malignant transformation of oral leukoplakia. We analysed the immunoexpression of BubR1, Mad2, Bub3, and Spindly proteins in 64 oral biopsies from 52 oral leukoplakias and 12 normal tissues. Univariate and multivariate analysis were performed to evaluate predictive factors for malignant transformation (MT). We observed that BubR1 and Mad2 were more highly expressed in high dysplasia grade lesions than in low grade or normal tissues (P<0.05). High expression of Spindly was significantly correlated with a high Ki-67 score (P=0.004). Six (11.5%) oral leukoplakias underwent malignant transformation. In univariate analysis, the binary dysplasia grade (high grade) (P<0.001) was associated with a higher risk of malignant transformation as well as high BubR1 (P<0.001) and high Mad2 (P=0.013) expression. In multivariate analysis, high expression of BubR1 and Mad2 when combined showed an increased risk for malignant transformation (P=0.013; HR of 4.6, 95% CI of 1.4-15.1). Our findings reveal that BubR1 and Mad2 were associated with an increased risk for malignant transformation independently of histological grade and could be potential and useful predictive risk markers of malignant transformation in oral leukoplakias.