Merck
  • Cancer-associated fibroblast-derived exosomal microRNA-24-3p enhances colon cancer cell resistance to MTX by down-regulating CDX2/HEPH axis.
Cancer-associated fibroblast-derived exosomal microRNA-24-3p enhances colon cancer cell resistance to MTX by down-regulating CDX2/HEPH axis.
Journal of cellular and molecular medicine (2021-02-24)
Hong-Wei Zhang, Yi Shi, Ji-Bin Liu, Hui-Min Wang, Pei-Yao Wang, Zhi-Jun Wu, Liu Li, Li-Peng Gu, Ping-Sheng Cao, Gao-Ren Wang, Yu-Shui Ma, Da Fu
ABSTRACT
MicroRNA-24-3p (miR-24-3p) has been implicated as a key promoter of chemotherapy resistance in numerous cancers. Meanwhile, cancer-associated fibroblasts (CAFs) can secret exosomes to transfer miRNAs, which mediate tumour development. However, little is known regarding the molecular mechanism of CAF-derived exosomal miR-24-3p in colon cancer (CC). Hence, this study intended to characterize the functional relevance of CAF-derived exosomal miR-24-3p in CC cell resistance to methotrexate (MTX). We identified differentially expressed HEPH, CDX2 and miR-24-3p in CC through bioinformatics analyses, and validated their expression in CC tissues and cells. The relationship among HEPH, CDX2 and miR-24-3p was verified using ChIP and dual-luciferase reporter gene assays. Exosomes were isolated from miR-24-3p inhibitor-treated CAFs (CAFs-exo/miR-24-3p inhibitor), which were used in combination with gain-of-function and loss-of-function experiments and MTX treatment. CCK-8, flow cytometry and colony formation assays were conducted to determine cell viability, apoptosis and colony formation, respectively. Based on the findings, CC tissues and cells presented with high expression of miR-24-3p and low expression of HEPH and CDX2. CDX2 was a target gene of miR-24-3p and could up-regulate HEPH. Under MTX treatment, overexpressed CDX2 or HEPH and down-regulated miR-24-3p reduced cell viability and colony formation and elevated cell apoptosis. Furthermore, miR-24-3p was transferred into CC cells via CAF-derived exosomes. CAF-derived exosomal miR-24-3p inhibitor diminished cell viability and colony formation and increased cell apoptosis in vitro and inhibited tumour growth in vivo under MTX treatment. Altogether, CAF-derived exosomal miR-24-3p accelerated resistance of CC cells to MTX by down-regulating CDX2/HEPH axis.
Sigma-Aldrich
Bicinchoninic Acid Kit for Protein Determination, for 200-1000 μg/ml protein