Merck
  • Effects of anesthetic MS-222 on stress and reproduction of South American silver catfish (Rhamdia quelen) males.
Effects of anesthetic MS-222 on stress and reproduction of South American silver catfish (Rhamdia quelen) males.
Animal reproduction science (2020-12-29)
Nathalia Dos Santos Teixeira, Lis Santos Marques, Rômulo Batista Rodrigues, Darlan Gusso, Gabriela Thais Pinheiro, Thales Lysakowski Flores Machado, Danilo Pedro Streit
ABSTRACT
Anesthesia is a common practice used in fish research and aquaculture. It is important to understand anesthetic effects on the animal and tissues of interest to ensure validity of data and to improve animal welfare in research and fish production endeavors. The production of some captive fish species is only possible by imposing artificial reproduction procedures, and manipulation of fish for these purposes is a stressor. The purpose of this study, therefore, was to evaluate effects of different concentrations (100, 200, and 300 mg/L) of the anesthetic MS-222 (tricaine methanesulfonate) on cortisol concentrations and effects on sperm quality in Rhamdia quelen. After hormonal induction of gamete production, 28 sexually mature males were randomly assigned to treatments, and milt and blood samples were collected. Anesthesia induction time, motility rate, sperm concentration and morphology, plasma cortisol concentrations, and reproductive hormone concentrations (testosterone, 17-α-hydroxyprogesterone, and estradiol) were evaluated. Sperm motility was greater in the control than 300 mg/L treatment group but did not differ among the control, 100, and 200 mg/L groups. The estradiol concentration was greater in non- anesthetized than anesthetized Rhamdia quelen, but plasma cortisol concentrations did not differ among treatment groups (182.50 ± 42.03 ng/mL). The anesthetic MS-222 at concentrations of 100, 200, and 300 mg/L did not inhibit the stress response due to handling of Rhamdia quelen males. In addition, treatment with MS-222 was not effective in inhibiting detrimental effects on sperm quality because this treatment was associated with impaired sperm motility and lesser concentrations of plasma estradiol.