Merck
  • Metabolic Effects of Late Dinner in Healthy Volunteers-A Randomized Crossover Clinical Trial.
Metabolic Effects of Late Dinner in Healthy Volunteers-A Randomized Crossover Clinical Trial.
The Journal of clinical endocrinology and metabolism (2020-06-12)
Chenjuan Gu, Nga Brereton, Amy Schweitzer, Matthew Cotter, Daisy Duan, Elisabet Børsheim, Robert R Wolfe, Luu V Pham, Vsevolod Y Polotsky, Jonathan C Jun
ABSTRACT
Consuming calories later in the day is associated with obesity and metabolic syndrome. We hypothesized that eating a late dinner alters substrate metabolism during sleep in a manner that promotes obesity. The objective of this work is to examine the impact of late dinner on nocturnal metabolism in healthy volunteers. This is a randomized crossover trial of late dinner (LD, 22:00) vs routine dinner (RD, 18:00), with a fixed sleep period (23:00-07:00) in a laboratory setting. Participants comprised 20 healthy volunteers (10 male, 10 female), age 26.0 ± 0.6 years, body mass index 23.2 ± 0.7 kg/m2, accustomed to a bedtime between 22:00 and 01:00. An isocaloric macronutrient diet was administered on both visits. Dinner (35% daily kcal, 50% carbohydrate, 35% fat) with an oral lipid tracer ([2H31] palmitate, 15 mg/kg) was given at 18:00 with RD and 22:00 with LD. Measurements included nocturnal and next-morning hourly plasma glucose, insulin, triglycerides, free fatty acids (FFAs), cortisol, dietary fatty acid oxidation, and overnight polysomnography. LD caused a 4-hour shift in the postprandial period, overlapping with the sleep phase. Independent of this shift, the postprandial period following LD was characterized by higher glucose, a triglyceride peak delay, and lower FFA and dietary fatty acid oxidation. LD did not affect sleep architecture, but increased plasma cortisol. These metabolic changes were most pronounced in habitual earlier sleepers determined by actigraphy monitoring. LD induces nocturnal glucose intolerance, and reduces fatty acid oxidation and mobilization, particularly in earlier sleepers. These effects might promote obesity if they recur chronically.