Merck
  • Potent and selective inhibition of human nitric oxide synthases. Selective inhibition of neuronal nitric oxide synthase by S-methyl-L-thiocitrulline and S-ethyl-L-thiocitrulline.
Potent and selective inhibition of human nitric oxide synthases. Selective inhibition of neuronal nitric oxide synthase by S-methyl-L-thiocitrulline and S-ethyl-L-thiocitrulline.
The Journal of biological chemistry (1994-10-28)
E S Furfine, M F Harmon, J E Paith, R G Knowles, M Salter, R J Kiff, C Duffy, R Hazelwood, J A Oplinger, E P Garvey
ABSTRACT
Potent and selective inhibition of neuronal nitric oxide synthase (nNOS) compared to endothelial NOS (eNOS) and inducible NOS (iNOS) may be useful to treat cerebral ischemia (stroke) and other neurodegenerative diseases. S-Methyl-L-thiocitrulline (Me-TC) and S-ethyl-L-thiocitrulline (Et-TC) inhibited the oxidation of L-arginine and the L-arginine-independent oxidation of NADPH by nNOS from human brain. Me-TC and Et-TC were slow, tight binding inhibitors of nNOS with second-order association rate constants (kon) of 2.6 x 10(5) M-1 s-1 and 1.3 x 10(5) M-1 s-1, respectively. The respective dissociation rate constants (koff) were 3 x 10(-4) s-1 and 0.7 x 10(-4) s-1. Thus, the Kd values calculated from koff/kon were 1.2 and 0.5 nM, respectively. L-Arginine was a competitive inhibitor of Me-TC and Et-TC binding with competition constant (Ks) values of 2.2 and 2.7 microM, respectively. The Km of nNOS for L-arginine was 1.6 microM. The active site concentration of nNOS was estimated by titration with Et-TC. Based on this active site concentration, a kcat of 0.4 s-1 for the oxidation of L-arginine, was calculated. Me-TC and Et-TC were less potent inhibitors of human iNOS (Ki values of 34 and 17 nM, respectively) and human eNOS (Ki values of 11 and 24 nM). Thus, Me-TC and Et-TC were 10- and 50-fold, respectively, more potent inhibitors of nNOS than eNOS. Furthermore, Me-TC was also 17-fold selective for rat nNOS in neuronal tissue compared to rat eNOS in vascular endothelium, suggesting that Me-TC may be selective for nNOS in vivo and therefore, may be therapeutically useful to treat neurodegenerative diseases.