Merck
  • Whole Genome Resequencing of Capsicum baccatum and Capsicum annuum to Discover Single Nucleotide Polymorphism Related to Powdery Mildew Resistance.
Whole Genome Resequencing of Capsicum baccatum and Capsicum annuum to Discover Single Nucleotide Polymorphism Related to Powdery Mildew Resistance.
Scientific reports (2018-03-28)
Yul-Kyun Ahn, Abinaya Manivannan, Sandeep Karna, Tae-Hwan Jun, Eun-Young Yang, Sena Choi, Jin-Hee Kim, Do-Sun Kim, Eun-Su Lee
ABSTRACT
The present study deals with genome wide identification of single-nucleotide polymorphism (SNP) markers related to powdery mildew (PM) resistance in two pepper varieties. Capsicum baccatum (PRH1- a PM resistant line) and Capsicum annuum (Saengryeg- a PM susceptible line), were resequenced to develop SNP markers. A total of 6,213,009 and 6,840,889 SNPs for PRH1 and Saengryeg respectively have been discovered. Among the SNPs, majority were classified as homozygous type SNPs, particularly in the resistant line. Moreover, the SNPs were differentially distributed among the chromosomes in both the resistant and susceptible lines. In total, 4,887,031 polymorphic SNP loci were identified between the two lines and 306,871 high-resolution melting (HRM) marker primer sets were designed. In order to understand the SNPs associated with the vital genes involved in diseases resistance and stress associated processes, chromosome-wise gene ontology analysis was performed. The results revealed the occurrence that SNPs related to diseases resistance genes were predominantly distributed in chromosome 4. In addition, 6281 SNPs associated with 46 resistance genes were identified. Among the lines, PRH1 consisted of maximum number of polymorphic SNPs related to NBS-LRR genes. The SNP markers were validated using HRM assay in 45 F4 populations and correlated with the phenotypic disease index.
Sigma-Aldrich
GenElute Plant Genomic DNA Miniprep Kit, sufficient for 70 purifications