Merck
  • The diverse roles of flavin coenzymes--nature's most versatile thespians.
The diverse roles of flavin coenzymes--nature's most versatile thespians.
The Journal of organic chemistry (2007-06-22)
Steven O Mansoorabadi, Christopher J Thibodeaux, Hung-wen Liu
ABSTRACT
Flavin coenzymes play a variety of roles in biological systems. This Perspective highlights the chemical versatility of flavins by reviewing research on five flavoenzymes that have been studied in our laboratory. Each of the enzymes discussed in this review [the acyl-CoA dehydrogenases (ACDs), CDP-6-deoxy-l-threo-d-glycero-4-hexulose-3-dehydrase reductase (E3), CDP-4-aceto-3,6-dideoxygalactose synthase (YerE), UDP-galactopyranose mutase (UGM), and type II isopentenyl diphosphate:dimethylallyl diphosphate isomerase (IDI-2)] utilizes flavin in a distinct role. In particular, the catalytic mechanisms of two of these enzymes, UGM and IDI-2, may involve novel flavin chemistry.
Sigma-Aldrich
Riboflavin 5′-monophosphate sodium salt hydrate, suitable for electrophoresis, suitable for acrylamide photopolymerization, ≥70%, powder
Sigma-Aldrich
Riboflavin 5′-monophosphate sodium salt hydrate, synthetic, ≥70% (HPLC)