- Synthesis of 2,3-Disubstituted 4-Ethoxycarbonyl-β-carbolin-1-ones: Application to the Synthesis of SL651498 and Its Analogue.
Synthesis of 2,3-Disubstituted 4-Ethoxycarbonyl-β-carbolin-1-ones: Application to the Synthesis of SL651498 and Its Analogue.
ACS omega (2020-07-09)
Ryoya Miura, Shinsuke Goto, Takahide Hashimoto, Iwao Hachiya
PMID32637839
ABSTRACT
Synthesis of 2,3-disubstituted 4-ethoxycarbonyl-β-carbolin-1-ones is developed using palladium-catalyzed intramolecular amination of 3-amino-4-(2-bromophenyl)-2-pyridones prepared by the conjugate addition reaction of diethyl 2-aminomalonate to alkynyl imines. The method was applied to the total syntheses of SL651498 exhibiting anxiolytic activity and its analogue.
MATERIALS