Merck
  • Priming exposures to lipopolysaccharides do not affect the induction of Polycomb target genes upon re-exposure.
Priming exposures to lipopolysaccharides do not affect the induction of Polycomb target genes upon re-exposure.
PloS one (2020-04-15)
Marco Geigges, Geethika Arekatla, Renato Paro
ABSTRACT
The Polycomb group (PcG) proteins are chromatin factors underlying the process of transcriptional memory to preserve developmental decisions and keep cellular identities. However, not only developmental signals need to be memorized and thus maintained during the life of an organism. For host protection against pathogens, also a memory of previous exposures to an immunogenic stimulus is crucial to mount a more protective immune response upon re-exposure. The antigen-specific adaptive immunity in vertebrates is an example of such a memory to previous immunogenic stimulation. Recently, adaptive characteristics were also attributed to innate immunity, which was classically seen to lack memory. However, the mechanistic details of an adaptive innate immune response are yet to be fully understood and chromatin-based epigenetic mechanisms seem to play an important role in this phenomenon. Possibly, PcG proteins can contribute to such an epigenetic innate immune memory. In this study, we analyzed whether the PcG system can mediate a transcriptional memory of exposure to lipopolysaccharides (LPS). To this end, various forms of LPS pre-treatment were applied to reporter cells and expression kinetics of PcG target genes were analyzed after a second LPS exposure. Neither single nor multiple LPS pre-treatment affected the induction of endogenous LPS-responsive transcripts upon re-exposure. Altogether, our extensive analyses did not provide any evidence for a PcG system-mediated memory of LPS stimulation.
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Schneider′s Insect Medium, With L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for insect cell culture