Merck
  • Sevelamer Use in End-Stage Kidney Disease (ESKD) Patients Associates with Poor Vitamin K Status and High Levels of Gut-Derived Uremic Toxins: A Drug-Bug Interaction?
Sevelamer Use in End-Stage Kidney Disease (ESKD) Patients Associates with Poor Vitamin K Status and High Levels of Gut-Derived Uremic Toxins: A Drug-Bug Interaction?
Toxins (2020-05-31)
Lu Dai, Björn K Meijers, Bert Bammens, Henriette de Loor, Leon J Schurgers, Abdul Rashid Qureshi, Peter Stenvinkel, Pieter Evenepoel
ABSTRACT
Gut microbial metabolism is not only an important source of uremic toxins but may also help to maintain the vitamin K stores of the host. We hypothesized that sevelamer therapy, a commonly used phosphate binder in patients with end-stage kidney disease (ESKD), associates with a disturbed gut microbial metabolism. Important representatives of gut-derived uremic toxins, including indoxyl sulfate (IndS), p-Cresyl sulfate (pCS), trimethylamine N-oxide (TMAO), phenylacetylglutamine (PAG) and non-phosphorylated, uncarboxylated matrix-Gla protein (dp-ucMGP; a marker of vitamin K status), were analyzed in blood samples from 423 patients (65% males, median age 54 years) with ESKD. Demographics and laboratory data were extracted from electronic files. Sevelamer users (n = 172, 41%) were characterized by higher phosphate, IndS, TMAO, PAG and dp-ucMGP levels compared to non-users. Sevelamer was significantly associated with increased IndS, PAG and dp-ucMGP levels, independent of age, sex, calcium-containing phosphate binder, cohort, phosphate, creatinine and dialysis vintage. High dp-ucMGP levels, reflecting vitamin K deficiency, were independently and positively associated with PAG and TMAO levels. Sevelamer therapy associates with an unfavorable gut microbial metabolism pattern. Although the observational design precludes causal inference, present findings implicate a disturbed microbial metabolism and vitamin K deficiency as potential trade-offs of sevelamer therapy.
Sigma-Aldrich
Phenylacetyl-L-glutamine, ≥95%