Merck
  • Embelin averts MPTP-induced dysfunction in mitochondrial bioenergetics and biogenesis via activation of SIRT1.
Embelin averts MPTP-induced dysfunction in mitochondrial bioenergetics and biogenesis via activation of SIRT1.
Biochimica et biophysica acta. Bioenergetics (2020-01-29)
Swetha Pavani Rao, Neelam Sharma, Shasi V Kalivendi
ABSTRACT
Parkinson's disease (PD) is a chronic neurodegenerative disease characterized by the death of dopamine neurons of Substantia nigra pars compacta (SNpc) leading to motor deficits. Amongst the mechanisms proposed, mitochondrial dysfunction, reduced complex-I and PGC1α levels were found to correlate with the pathology of PD. As embelin is a natural product with structural resemblance to ubiquinone, exhibits mitochondrial uncoupling and antioxidant effects, in the present study, we sought to examine its role in the mechanisms mediating PD. Results indicate that embelin protects from MPP+-induced oxidative stress and apoptosis in a time and dose-dependent manner in N27 dopaminergic cells. Cells treated with embelin exhibited increased levels of pAMPK, SIRT1 and PGC1α leading to enhanced mitochondrial biogenesis. Though treatment of cells with MPP+ also increased pAMPK levels, but, SIRT1 and PGC1α levels decreased substantially, possibly due to the block in the mitochondrial electron transport chain and reduced NAD/NADH levels. The mitochondrial uncoupling effects of embelin leading to increased NAD/NADH levels followed by enhanced SIRT1, PGC1α and mitochondrial biogenesis were found to confer embelin mediated protection as treatment of cells with SIRT1 inhibitor or siRNA nullified this effect. Embelin (10 mg/kg) also conferred protection in vivo in MPTP mouse model of PD, wherein, MPTP-induced loss of TH staining, reduced striatal dopamine and markers of mitochondrial biogenesis pathway were averted by embelin.
Sigma-Aldrich
Embelin, ≥98% (HPLC), powder