Merck
  • Ethylenediamine-N,N'-Disuccinic Acid (EDDS)-Enhanced Flushing Optimization for Contaminated Agricultural Soil Remediation and Assessment of Prospective Cu and Zn Transport.
Ethylenediamine-N,N'-Disuccinic Acid (EDDS)-Enhanced Flushing Optimization for Contaminated Agricultural Soil Remediation and Assessment of Prospective Cu and Zn Transport.
International journal of environmental research and public health (2018-03-23)
Marco Race, Alberto Ferraro, Massimiliano Fabbricino, Agostino La Marca, Antonio Panico, Danilo Spasiano, Alice Tognacchini, Francesco Pirozzi
ABSTRACT
This paper presents the results of an experimental study aimed at investigating the effect of operative parameters on the efficiency of a soil flushing process, conducted on real contaminated soil containing high amounts of Cu and Zn. Soil flushing tests were carried out with Ethylenediamine-N,N'-disuccinic acid (EDDS) as a flushing agent due to its high biodegradability and environmentally friendly characteristics. Process parameters such as Empty-Bed Contact Time (EBCT) and EDDS solution molarity were varied from 21-33 h and from 0.36-3.6 mM, respectively. Effects on the mobility of cations such as Fe and Mn were also investigated. Results showed that very high performances can be obtained at [EDDS] = 3.6 mM and EBCT = 33 h. In these conditions, in fact, the amount of removed Cu was 53%, and the amount of removed Zn was 46%. Metal distribution at different depths from the top surface revealed that Cu has higher mobility than Zn. The process results were strongly dependent on the exchange of metals due to the different stability constants of the EDDS complexes. Finally, results from a comparative study showed that soil washing treatment reached the same removal efficiency of the flushing process in a shorter time but required a larger amount of the EDDS solution.
Sigma-Aldrich
Ethylenediamine-N,N′-disuccinic acid trisodium salt solution, ~35% in H2O