Merck
  • Identification of α-Tomatine 23-Hydroxylase Involved in the Detoxification of a Bitter Glycoalkaloid.
Identification of α-Tomatine 23-Hydroxylase Involved in the Detoxification of a Bitter Glycoalkaloid.
Plant & cell physiology (2019-12-10)
Masaru Nakayasu, Ryota Akiyama, Midori Kobayashi, Hyoung Jae Lee, Takashi Kawasaki, Bunta Watanabe, Shingo Urakawa, Junpei Kato, Yukihiro Sugimoto, Yoko Iijima, Kazuki Saito, Toshiya Muranaka, Naoyuki Umemoto, Masaharu Mizutani
ABSTRACT
Tomato plants (Solanum lycopersicum) contain steroidal glycoalkaloid α-tomatine, which functions as a chemical barrier to pathogens and predators. α-Tomatine accumulates in all tissues and at particularly high levels in leaves and immature green fruits. The compound is toxic and causes a bitter taste, but its presence decreases through metabolic conversion to nontoxic esculeoside A during fruit ripening. This study identifies the gene encoding a 23-hydroxylase of α-tomatine, which is a key to this process. Some 2-oxoglutarate-dependent dioxygenases were selected as candidates for the metabolic enzyme, and Solyc02g062460, designated Sl23DOX, was found to encode α-tomatine 23-hydroxylase. Biochemical analysis of the recombinant Sl23DOX protein demonstrated that it catalyzes the 23-hydroxylation of α-tomatine and the product spontaneously isomerizes to neorickiioside B, which is an intermediate in α-tomatine metabolism that appears during ripening. Leaves of transgenic tomato plants overexpressing Sl23DOX accumulated not only neorickiioside B but also another intermediate, lycoperoside C (23-O-acetylated neorickiioside B). Furthermore, the ripe fruits of Sl23DOX-silenced transgenic tomato plants contained lower levels of esculeoside A but substantially accumulated α-tomatine. Thus, Sl23DOX functions as α-tomatine 23-hydroxylase during the metabolic processing of toxic α-tomatine in tomato fruit ripening and is a key enzyme in the domestication of cultivated tomatoes.
Sigma-Aldrich
Tomatidine hydrochloride, ≥85%