Merck
  • Supramolecular Sandwiches: Halogen-Bonded Coformers Direct [2+2] Photoreactivity in Two-Component Cocrystals.
Supramolecular Sandwiches: Halogen-Bonded Coformers Direct [2+2] Photoreactivity in Two-Component Cocrystals.
Molecules (Basel, Switzerland) (2020-02-23)
Jay Quentin, Dale C Swenson, Leonard R MacGillivray
ABSTRACT
The halogen-bond (X-bond) donors 1,3- and 1,4-diiodotetrafluorobenzene (1,3-di-I-tFb and 1,4-di-I-tFb, respectively) form cocrystals with trans-1,2-bis(2-pyridyl)ethylene (2,2'-bpe) assembled by N···I X-bonds. In each cocrystal, 2(1,3-di-I-tFb)·2(2,2'-bpe) and (1,4-di-I-tFb)·(2,2'-bpe), the donor molecules support the C=C bonds of 2,2'-bpe to undergo an intermolecular [2+2] photodimerization. UV irradiation of each cocrystal resulted in stereospecific and quantitative conversion of 2,2'-bpe to rctt-tetrakis(2-pyridyl)cyclobutane (2,2'-tpcb). In each case, the reactivity occurs via face-to-face π-stacked columns wherein nearest-neighbor pairs of 2,2'-bpe molecules lie sandwiched between X-bond donor molecules. Nearest-neighbor C=C bonds are stacked criss-crossed in both cocrystals. The reactivity was ascribed to the olefins undergoing pedal-like motion in the solid state. The stereochemistry of 2,2'-tpcb is confirmed in cocrystals 2(1,3-di-I-tFb)·(2,2'-tpcb) and (1,4-di-I-tFb)·(2,2'-tpcb).
Sigma-Aldrich
1,4-Diiodotetrafluorobenzene, 98%