Merck
  • Genome-wide Screening Identifies SFMBT1 as an Oncogenic Driver in Cancer with VHL Loss.
Genome-wide Screening Identifies SFMBT1 as an Oncogenic Driver in Cancer with VHL Loss.
Molecular cell (2020-02-06)
Xijuan Liu, Jeremy M Simon, Haibiao Xie, Lianxin Hu, Jun Wang, Giada Zurlo, Cheng Fan, Travis S Ptacek, Laura Herring, Xianming Tan, Mingjie Li, Albert S Baldwin, William Y Kim, Tao Wu, Marc W Kirschner, Kan Gong, Qing Zhang
ABSTRACT
von Hippel-Lindau (VHL) is a critical tumor suppressor in clear cell renal cell carcinomas (ccRCCs). It is important to identify additional therapeutic targets in ccRCC downstream of VHL loss besides hypoxia-inducible factor 2α (HIF2α). By performing a genome-wide screen, we identified Scm-like with four malignant brain tumor domains 1 (SFMBT1) as a candidate pVHL target. SFMBT1 was considered to be a transcriptional repressor but its role in cancer remains unclear. ccRCC patients with VHL loss-of-function mutations displayed elevated SFMBT1 protein levels. SFMBT1 hydroxylation on Proline residue 651 by EglN1 mediated its ubiquitination and degradation governed by pVHL. Depletion of SFMBT1 abolished ccRCC cell proliferation in vitro and inhibited orthotopic tumor growth in vivo. Integrated analyses of ChIP-seq, RNA-seq, and patient prognosis identified sphingosine kinase 1 (SPHK1) as a key SFMBT1 target gene contributing to its oncogenic phenotype. Therefore, the pVHL-SFMBT1-SPHK1 axis serves as a potential therapeutic avenue for ccRCC.
Sigma-Aldrich
Anti-SFMBT1 antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Deferoxamine mesylate salt, powder, ≥92.5% (TLC)
Sigma-Aldrich
Catalase from bovine liver, aqueous suspension, 40,000-60,000 units/mg protein (E1%/405)
Sigma-Aldrich
Anti-SFMBT1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody
Roche
Anti-HA Affinity Matrix, from rat IgG1
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution