Merck
  • Orthogonal Supramolecular Assembly Triggered by Inclusion and Exclusion Interactions with Cucurbit[7]uril for Photocatalytic H2 Evolution.
Orthogonal Supramolecular Assembly Triggered by Inclusion and Exclusion Interactions with Cucurbit[7]uril for Photocatalytic H2 Evolution.
ChemSusChem (2019-11-05)
Dengmeng Song, Bo Li, Xin Li, Xuzhuo Sun, Jun Li, Chengbo Li, Tongyu Xu, Yong Zhu, Fei Li, Ning Wang
ABSTRACT
The fabrication of efficient and convenient photocatalytic H2 evolution systems is a fascinating research topic in the field of solar energy conversion. A ternary self-assembled photocatalytic H2 evolution system was fabricated through supramolecular host-guest chemistry. The system consisted of the H2 evolution catalyst [Co(dmgH)2 (4-ppy)2 ]NO3 (1; dmgH2 =dimethylglyoxime, 4-ppy=4-phenylpyridine) and the photosensitizer Eosin Y (EY) assembled with the macrocyclic compound cucurbit[7]uril (CB[7]) to form the 1@CB[7]/EY complex through inclusion and exclusion interactions, respectively. The synchronous self-assembly drives an orthogonal arrangement of the 1@CB[7]/EY system. The inclusion complex 1@CB[7] was successfully characterized by 1 H NMR spectroscopy and single-crystal XRD. The exclusion process of CB[7] with EY was identified by NMR titration and the optimized geometry of the exclusion structure was determined by DFT calculations. The use of CB[7] resulted in a 6-fold increase in turnover number, a 3-fold increase in turnover frequency, and a 3-fold extension of lifetime for photocatalytic H2 evolution as compared with the system in the absence of CB[7]. The improvement of the light-driven H2 evolution activity was ascribed to the ability of CB[7] to link the photosensitizer and catalyst.
Sigma-Aldrich
4-Phenylpyridine, 97%