Merck
  • Harmol induces apoptosis by caspase-8 activation independently of Fas/Fas ligand interaction in human lung carcinoma H596 cells.
Harmol induces apoptosis by caspase-8 activation independently of Fas/Fas ligand interaction in human lung carcinoma H596 cells.
Anti-cancer drugs (2009-03-26)
Akihisa Abe, Hiroyuki Yamada
ABSTRACT
The beta-carboline alkaloids are naturally existing plant substances. It is known that these alkaloids have a wide spectrum of neuropharmacological, psychopharmacological, and antitumor effects. Therefore, they have been traditionally used in oriental medicine for the treatment of various diseases including cancers and malaria. In this study, harmol and harmalol, which are beta-carboline alkaloids, were examined for their antitumor effect on human lung carcinoma cell lines, and structure-activity relationship was also investigated. H596, H226, and A549 cells were treated with harmol and harmalol, respectively. Apoptosis was induced by harmol only in H596 cells. In contrast, harmalol had negligible cytotoxicity in three cell lines. Harmol induced caspase-3, caspase-8, and caspase-9 activities and caspase-3 activities accompanied by cleavage of poly-(ADP-ribose)-polymerase. Furthermore, harmol treatment decreased the native Bid protein, and induced the release of cytochrome c from mitochondria to cytosol. The apoptosis induced by harmol was completely inhibited by caspase-8 inhibitor and partially inhibited by caspase-9 inhibitor. The antagonistic antibody ZB4 blocked Fas ligand-induced apoptosis, but had no effect on harmol-induced apoptosis. Harmol had no significant effect on the expression of Fas. In conclusion, our results showed that the harmol could cause apoptosis-inducing effects in human lung H596 cells through caspase-8-dependent pathway but independent of Fas/Fas ligand interaction.