Merck
  • DNA damage-induced cellular senescence is sufficient to suppress tumorigenesis: a mouse model.
DNA damage-induced cellular senescence is sufficient to suppress tumorigenesis: a mouse model.
The Journal of experimental medicine (2007-05-31)
Thang Van Nguyen, Nahum Puebla-Osorio, Hui Pang, Melanie E Dujka, Chengming Zhu
ABSTRACT
Tumor suppressor p53-dependent apoptosis is critical in suppressing tumorigenesis. Previously, we reported that DNA double-strand breaks (DSBs) at the V(D)J recombination loci induced genomic instability in the developing lymphocytes of nonhomologous end-joining (NHEJ)-deficient, p53-deficient mice, which led to rapid lymphomagenesis. To test the ability of p53-dependent cell cycle arrest to suppress tumorigenesis in the absence of apoptosis in vivo, we crossbred NHEJ-deficient mice into a mutant p53R172P background; these mice have defects in apoptosis induction, but not cell cycle arrest. These double-mutant mice survived longer than NHEJ/p53 double-null mice and, remarkably, were completely tumor free. We detected accumulation of aberrant V(D)J recombination-related DSBs at the T cell receptor (TCR) locus, and high expression levels of both mutant p53 and cell cycle checkpoint protein p21, but not the apoptotic protein p53-upregulated modulator of apoptosis. In addition, a substantial number of senescent cells were observed among both thymocytes and bone marrow cells. Cytogenetic studies revealed euploidy and limited chromosomal breaks in these lymphoid cells. The results indicate that precursor lymphocytes, which normally possess a high proliferation potential, are able to withdraw from the cell cycle and undergo senescence in response to the persistence of DSBs in a p53-p21-dependent pathway; this is sufficient to inhibit oncogenic chromosomal abnormality and suppress tumorigenesis.
Sigma-Aldrich
Anti-HP1γ Antibody, clone 42s2, clone 42s2, Upstate®, from mouse