Merck
  • Serotonin via 5-HT1B and 5-HT2B receptors stimulates anion secretion in the rat epididymal epithelium.
Serotonin via 5-HT1B and 5-HT2B receptors stimulates anion secretion in the rat epididymal epithelium.
The Journal of physiology (1999-08-24)
G P Leung, S L Dun, N J Dun, P Y Wong
ABSTRACT
1. The short-circuit current (Isc) technique was used to study the role of 5-hydroxytryptamine (5-HT) in the regulation of anion secretion in cultured rat cauda epididymal epithelia. 2. 5-HT, the 5-HT1B-selective agonist 5-nonyloxytryptamine (5-NOT) and the 5-HT2B-selective agonist alpha-methyl-5-hydroxytryptamine (alpha-methyl-5-HT) added basolaterally stimulated Isc in a dose-dependent manner with EC50 values of 0.4, 20 and 0.3 microM, respectively. No other agonists for 5-HT receptors had any effect. 3. The pattern of responses to 5-HT was biphasic. Pretreating the tissues with the 5-HT1B-selective antagonist isamoltane (200 microM) and the 5-HT2B-selective antagonist rauwolscine (200 microM) inhibited the rapid transient phase by 55 and 45 %, whereas the sustained phase could only be blocked by rauwolscine. 4. Removal of chloride or bicarbonate or both from the normal Krebs-Henseleit solution reduced the responses to 5-HT, 5-NOT and alpha-methyl-5-HT to varying degrees. The results suggest that 5-HT1B- and 5-HT2B-mediated responses were mainly due to chloride and bicarbonate secretion, respectively. 5. Manipulation of the cAMP and Ca2+ signal transduction pathways with chemical agents provided evidence that the responses to 5-HT were mediated through cAMP. 6. Piroxicam pretreatment abolished the Isc response to alpha-methyl-5-HT but not to 5-NOT, indicating that the 5-HT2B-mediated response, but not the 5-HT1B-mediated response, is dependent on prostaglandin synthesis. 7. Immunohistochemical studies showed that 5-HT-like immunoreactivity was detected in nerve fibres and in small granular cells surrounding the epididymal tubules. 8. It is suggested that the 5-HT released from serotonergic nerve endings and/or from mast cells regulates electrolyte and fluid secretion in the epididymis.
Sigma-Aldrich
α-Methylserotonin maleate salt, ≥98% (HPLC)