- Inhibition of USP4 attenuates pathological scarring by downregulation of the TGF‑β/Smad signaling pathway.
Inhibition of USP4 attenuates pathological scarring by downregulation of the TGF‑β/Smad signaling pathway.
Pathological scarring is a result of the hypertrophy of scar tissue during tissue repair following trauma. The aim of the present study was to assess the effect of ubiquitin‑specific protease 4 (USP4) silencing on pathological scarring, and to evaluate the mechanistic basis for the effect. An MTT assay was used to assess cell viability. Immunoprecipitation (IP) was used to determine ubiquitination levels of the TGF‑β receptor (TβR)I and Smad7. Tumor formation was assessed by injecting keloid fibroblasts. Hematoxylin and eosin staining was used to detect pathological changes in tumor tissue. Reverse transcription quantitative polymerase chain reaction and western blot analysis assays were used to evaluate the expression levels of TβRI and Smad7. Compared with the untreated control animals, cell viability and the expression of TβRI and Smad7 increased significantly in animals treated with TGF‑β. Short hairpin RNA for USP4 (shUSP4) decreased the cell viability of negative control cells, TGF‑β‑induced cellular proliferation, and the expression of TβRI and Smad7. IP experiments indicated that the ubiquitination level of TβRI was decreased following USP4 silencing. There was no remarkable difference in the structure of scar tissue among the various animal groups at 14 days following treatment, while the necrotic area of the scar tissue in the shUSP4 and vialinin A (USP inhibitor)‑treated animals increased significantly at the 28th and 42nd day compared with the control animals. At days 14, 28 and 42, the expression levels of TβRI and Smad7 in the shUSP4 and vialinin A‑treated animals were significantly decreased compared with the control animals (P<0.05). In summary, interference with or inhibition of USP4 prevented the activity of the TGF‑β/Smad pathway signaling and inhibited the formation of pathological scars.