- Inhibition of atrial wall stretch-induced cardiac hormone secretion by lavendustin A, a potent tyrosine kinase inhibitor.
Inhibition of atrial wall stretch-induced cardiac hormone secretion by lavendustin A, a potent tyrosine kinase inhibitor.
The cellular processes linking mechanical wall stretch to atrial natriuretic peptide (ANP) and B-type natriuretic peptide (BNP) secretion from the heart are unclear. In the present study, a paced perfused rat heart preparation was used to study the signaling mechanisms of atrial wall stretch-induced secretion of ANP and BNP. Vehicle or drugs were infused into the perfusate for 40 min and right atrial wall stretch was superimposed for 10 min after 25-min drug infusions by elevating the level of the pulmonary artery cannula tip. Lavendustin A, a potent inhibitor of protein tyrosine kinases, at the concentrations of 0.5 and 1.3 microM decreased atrial wall stretch-induced ANP secretion (53% and 68%, respectively, P < 0.001) in the perfused rat heart preparation, whereas no difference in the hemodynamic variables (heart rate, contractile force and perfusion pressure) were noted between groups. Lavendustin A also completely abolished the wall stretch-induced secretion of BNP. Several other protein kinase inhibitors including staurosporine (protein kinase C inhibitor), ML-9 (myosin light chain kinase inhibitor), KN-62 (Ca2+/calmodulin-dependent protein kinase II inhibitor) and H-89 (protein kinase A inhibitor) had no significant effect on atrial wall stretch-stimulated ANP secretion. In a separate series of experiments, in which the right atria were stretched for 2 h, administration of lavendustin A (1 microM) but not staurosporine (30 nM) significantly decreased sustained wall stretch-induced ANP secretion. Okadaic acid, a potent protein phosphatase A2 (PPA2) and PP1 inhibitor, at the concentration of 100 nM had no effect on basal ANP secretion but significantly accelerated the ANP secretory response to atrial wall stretch (P < 0.05). In conclusion, the findings that inhibitors of protein tyrosine kinase and protein phosphatase selectively modulated atrial wall stretch-induced ANP secretion suggest a new mechanism involving endogenous protein tyrosine activity in the regulation of natriuretic peptide exocytosis from cardiac myocytes.