Merck
  • Melatonin: The smart molecule that differentially modulates autophagy in tumor and normal placental cells.
Melatonin: The smart molecule that differentially modulates autophagy in tumor and normal placental cells.
PloS one (2019-01-11)
Lucas Sagrillo-Fagundes, Josianne Bienvenue-Pariseault, Cathy Vaillancourt
ABSTRACT
Melatonin has protective roles in normal cells and cytotoxic actions in cancer cells, with effects involving autophagy and nuclear factor (erythroid-derived 2)-like 2 (Nrf2) transcription factor pathways. Hypoxia/reoxygenation (H/R) induces oxidative damage and apoptosis. These consequences activate autophagy, which degrades damaged cellular content, as well as activates Nrf2 the nuclear factor (erythroid-derived 2)-like 2 (Nrf2) transcription factor, and thereby the expression of protective genes. Melatonin has protective roles in normal cells and cytotoxic actions in cancer cells, with effects involving autophagy and Nrf2 pathways. The current study shows melatonin to differentially modulate autophagy and Nrf2 pathways in tumor and normal placental cells exposed to H/R. BeWo, a human placental choriocarcinoma cell line, and primary villous cytotrophoblasts isolated from normal term placenta, were maintained in normoxia (8% O2) for 24 h or exposed to hypoxia (0.5% of O2 for 4 h) followed by 20 h of normoxia, creating a situation of H/R, in the presence or absence of 1 mM melatonin. Melatonin induced a 7-fold increase in the activation of 5' adenosine monophosphate-activated protein kinase (AMPK)α, an upstream modulator of autophagy, rising to a 16-fold increase in BeWo cells co-exposed to H/R and melatonin, compared to controls. H/R induced autophagosome formation via the increased expression of Beclin-1 (by 94%) and ATG7 (by 97%) in BeWo cells. Moreover, H/R also induced autophagic activity, indicated by the by the 630% increase in P62, and increased Nrf2 by 314% in BeWo cells. In H/R conditions, melatonin reduced autophagic activity by 74% and Nrf2 expression activation by 300%, leading to BeWo cell apoptosis. In contrast, In human primary villous cytotrophoblasts, H/R induced autophagy and Nrf2, which melatonin further potentiated, thereby affording protection against H/R. This study demonstrates that melatonin differentially modulates autophagy and the Nrf2 pathway in normal vs. tumor trophoblast cells, being cytoprotective in normal cells whilst increasing apoptosis in tumoral trophoblast cells.
Sigma-Aldrich
Donkey Anti-Rabbit IgG Antibody, HRP conjugate, Species Adsorbed, Chemicon®, from donkey
Sigma-Aldrich
Donkey Anti-Goat IgG Antibody, HRP conjugate, Species Adsorbed, Chemicon®, from donkey
Sigma-Aldrich
Donkey Anti-Mouse IgG Antibody, HRP conjugate, Species Adsorbed, Chemicon®, from donkey
Sigma-Aldrich
Anti-PP2A Antibody, C subunit, clone 1D6, clone 1D6, Upstate®, from mouse
Sigma-Aldrich
Anti-LC3B antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution