Merck
  • Sequencing of Porphyromonas gingivalis from saliva in patients with periodontitis and type 2 diabetes mellitus.
Sequencing of Porphyromonas gingivalis from saliva in patients with periodontitis and type 2 diabetes mellitus.
Indian journal of medical microbiology (2019-08-20)
Preethi Radhakrishnan, Rubini Anbalagan, Ramya Barani, Monika Mani, Krishna G Seshadri, Padma Srikanth
ABSTRACT
Porphyromonas gingivalis is a major periodontal pathogen. Saliva is the most easy, non-invasive microbiological sample for detection of periodontal pathogens. A prospective study on 37 diabetic patients was grouped into well-controlled diabetes with/without periodontitis and uncontrolled diabetic with periodontitis. PCR and sequencing of P. gingivalis was performed in saliva samples. DNA was extracted from saliva using Triton X-100 and 16s rRNA gene (404 bp) was amplified by polymerase chain reaction. DNA sequencing was performed for two samples. P. gingivalis was detected in 27.03% (n = 10), of which 30% (n = 9) were diabetic with periodontal disease and 14.3% (n = 1) were diabetic without periodontal disease. The percentage of poor oral hygiene was 50% and 20% in uncontrolled and controlled glycaemic patients, respectively. DNA sequencing of two samples showed 100% identity with the sequences in the GenBank database (Gen Bank accession no: KX640913-KX640914). Type 2 diabetes mellitus and periodontitis are interlinked. Early detection of P. gingivalis and appropriate treatment with doxycycline will also assist in controlling the glycaemic status.